siRNA Details
| Gene Symbol: Fam131b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113327.2 | NP_001106798.1 | 6 | 843 | ||
| NM_001286584.1 | NP_001273513.1 | 7 | 819 | |||
| NM_029528.5 | NP_083804.3 | 7 | 879 | |||
| GenBank mRNA | AK018206.1 | - | 7 | 818 | ||
| AK018325.1 | - | 7 | 825 | |||
| AK047763.1 | - | 7 | 805 | |||
| AK158869.1 | - | 7 | 772 | |||
| AK163654.1 | - | 6 | 764 | |||
| BC115595.1 | - | 6 | 609 | |||
| BC115596.1 | - | 6 | 609 | |||

Fam131b
Genomic Map