siRNA Details
| Gene Symbol: Fam131b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113327.2 | NP_001106798.1 | 3, 4 | 412 | ||
| NM_001286584.1 | NP_001273513.1 | 4, 5 | 388 | |||
| NM_029528.5 | NP_083804.3 | 4, 5 | 448 | |||
| GenBank mRNA | AK018206.1 | - | 4, 5 | 387 | ||
| AK018325.1 | - | 4, 5 | 394 | |||
| AK047763.1 | - | 4, 5 | 374 | |||
| AK158869.1 | - | 4, 5 | 341 | |||
| AK163654.1 | - | 3, 4 | 333 | |||
| BC115595.1 | - | 3, 4 | 178 | |||
| BC115596.1 | - | 3, 4 | 178 | |||

Fam131b
Genomic Map