siRNA Details
| Gene Symbol: Slc35d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029529.3 | NP_083805.1 | 2 | 1622 | ||
| GenBank mRNA | AK018094.1 | - | 1 | 404 | ||
| AK042761.1 | - | 2 | 1374 | |||
| AK044973.1 | - | 2 | 1359 | |||
| AK047610.1 | - | 2 | 1623 | |||
| BC139194.1 | - | 2 | 1452 | 8862216 | ||

Slc35d3
Genomic Map