siRNA Details
| Gene Symbol: Coa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198006.4 | NP_932123.3 | 3 | 409 | ||
| GenBank mRNA | AK018255.1 | - | 3 | 285 | ||
| AK050432.1 | - | 3 | 315 | |||
| AK166651.1 | - | 3 | 339 | |||
| AK169639.1 | - | 3 | 351 | |||
| AK177444.1 | - | 3 | 230 | |||
| AK218336.1 | - | 3 | 230 | |||
| BB860009.1 | - | 3 | 360 | |||
| BB863739.1 | - | 3 | 352 | |||
| BC002137.1 | - | 3 | 316 | 3484305 | ||

Coa5
Genomic Map