siRNA Details
| Gene Symbol: Gpr165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029536.3 | NP_083812.1 | 1, 2 | 986 | ||
| GenBank mRNA | AK018327.1 | - | 1, 2 | 1014 | ||
| AK034479.1 | - | 1, 2 | 930 | |||
| AK038566.1 | - | 1, 2 | 258 | |||
| AK042723.1 | - | 1, 2 | 963 | |||
| AK045935.1 | - | 1, 2 | 942 | |||
| BC113184.1 | - | 1, 2 | 837 | 40061761 | ||
| BC113791.1 | - | 1, 2 | 837 | 40061764 | ||
More Information
Set Membership: |
|

Gpr165
Genomic Map