siRNA Details
| Gene Symbol: Agbl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289656.1 | NP_001276585.1 | 10, 11 | 2326 | ||
| NM_001289657.1 | NP_001276586.1 | 9, 10 | 2300 | |||
| NM_178630.4 | NP_848745.2 | 9, 10 | 2311 | |||
| GenBank mRNA | AK029793.1 | - | 10, 11 | 2327 | ||
| AK029798.1 | - | 9, 10 | 1884 | |||
| DQ867031.1 | - | 9, 10 | 2312 | |||
| DQ867032.1 | - | 9, 10 | 2312 | |||

Agbl3
Genomic Map