siRNA Details
| Gene Symbol: Agbl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289656.1 | NP_001276585.1 | 9 | 2166 | ||
| NM_001289657.1 | NP_001276586.1 | 8 | 2140 | |||
| NM_178630.4 | NP_848745.2 | 8 | 2151 | |||
| GenBank mRNA | AK029793.1 | - | 9 | 2167 | ||
| AK029798.1 | - | 8 | 1724 | |||
| AK030378.1 | - | 8 | 1709 | |||
| AK054066.1 | - | 7 | 3171 | |||
| BC139065.1 | - | 8 | 1603 | 8862087 | ||
| DQ867031.1 | - | 8 | 2152 | |||
| DQ867032.1 | - | 8 | 2152 | |||

Agbl3
Genomic Map