siRNA Details
| Gene Symbol: Grhpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080289.1 | NP_525028.1 | 6 | 568 | ||
| GenBank mRNA | AK004307.1 | - | 6 | 584 | ||
| AK018293.1 | - | 6 | 582 | |||
| AK168351.1 | - | 6 | 565 | |||
| AK172201.1 | - | 6 | 536 | |||
| AY113690.1 | - | 6 | 542 | |||
| BC010194.1 | - | 6 | 568 | 3966541 | ||

Grhpr
Genomic Map