siRNA Details
| Gene Symbol: Ercc6l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013608.2 | NP_001013626.2 | 2 | 523 | ||
| NM_023507.3 | NP_075996.2 | 2 | 509 | |||
| GenBank mRNA | AF217319.1 | - | 1 | 326 | ||
| AK002307.1 | - | 2 | 510 | |||
| AK035054.1 | - | 2 | 475 | |||
| AK049874.1 | - | 2 | 835 | |||
| AK050437.1 | - | 1 | 359 | |||
| BU148612.1 | - | 2 | 541 | |||

Ercc6l2
Genomic Map