siRNA Details
| Gene Symbol: Gstk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029555.2 | NP_083831.1 | 2, 3 | 239 | ||
| GenBank mRNA | AK002661.1 | - | 2, 3 | 237 | ||
| AY279096.1 | - | 2, 3 | 152 | |||
| BC038151.1 | - | 2, 3 | 166 | |||
| BC051507.1 | - | 2, 3 | 203 | |||
| BC058163.1 | - | 2, 3 | 172 | 5100572 | ||

Gstk1
Genomic Map