siRNA Details
| Gene Symbol: Asb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029569.3 | NP_083845.1 | 5 | 662 | ||
| GenBank mRNA | AF398966.1 | - | 5 | 573 | ||
| AK003770.1 | - | 5 | 663 | |||
| BC108405.1 | - | 5 | 577 | 30288598 | ||
| BC120851.1 | - | 5 | 628 | 40129774 | ||
| BC132007.1 | - | 5 | 628 | 40130284 | ||

Asb5
Genomic Map