siRNA Details
| Gene Symbol: Atp11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029570.3 | NP_083846.2 | 24, 25 | 3095 | ||
| GenBank mRNA | AK043943.1 | - | 13, 14 | 1744 | ||
| AK129248.1 | - | 24, 25 | 3087 | |||
| BC026986.1 | - | 9, 10 | 1164 | |||
| BC076603.1 | - | 24, 25 | 2965 | 30629594 | ||

Atp11b
Genomic Map