siRNA Details
| Gene Symbol: Rab1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029576.3 | NP_083852.1 | 3 | 182 | ||
| GenBank mRNA | AB232584.1 | - | 2 | 124 | ||
| AK003609.1 | - | 3 | 183 | |||
| AK042020.1 | - | 3 | 184 | |||
| AK156726.1 | - | 3 | 205 | |||
| AK208578.1 | - | 1 | 46 | |||
| AK215506.1 | - | 205 | ||||
| BC016408.1 | - | 3 | 177 | 4242206 | ||

Rab1b
Genomic Map