siRNA Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029746.3 | NP_084022.2 | 9 | 1043 | ||
| GenBank mRNA | AK089920.1 | - | 9 | 1027 | ||
| AK157348.1 | - | 9 | 1005 | |||
| AK169298.1 | - | 9 | 1013 | |||
| AK171726.1 | - | 9 | 962 | |||
| BC011435.1 | - | 9 | 983 | 4010102 | ||
| CX205121.1 | - | 3 | 252 | |||

Cog2
Genomic Map