siRNA Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029746.3 | NP_084022.2 | 1 | 158 | ||
| GenBank mRNA | AK012236.1 | - | 1 | 92 | ||
| AK089920.1 | - | 1 | 142 | |||
| AK157348.1 | - | 1 | 120 | |||
| AK169298.1 | - | 1 | 128 | |||
| AK171726.1 | - | 1 | 77 | |||
| BC011435.1 | - | 1 | 98 | 4010102 | ||
| CJ093818.1 | - | 1 | 158 | |||

Cog2
Genomic Map