siRNA Details
| Gene Symbol: Rab2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172601.3 | NP_766189.1 | 5 | 491 | ||
| GenBank mRNA | AB232586.1 | - | 5 | 337 | ||
| AK005230.1 | - | 5 | 394 | |||
| AK038393.1 | - | 5 | 426 | |||
| AK044202.1 | - | 5 | 368 | |||
| AK160249.1 | - | 5 | 642 | |||
| AK160351.1 | - | 5 | 357 | |||
| AK169657.1 | - | 5 | 435 | |||
| BC033312.1 | - | 5 | 388 | 4481516 | ||
| BC046334.1 | - | 5 | 433 | 6492838 | ||
| BC054719.1 | - | 5 | 424 | |||
| BC061513.1 | - | 1 | 440 | |||

Rab2b
Genomic Map