siRNA Details
| Gene Symbol: Ankrd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023816.2 | NP_076305.2 | 17 | 2910 | ||
| GenBank mRNA | AK005775.1 | - | 2 | 715 | ||
| AK132751.1 | - | 17 | 2909 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd36| Gene Symbol: Ankrd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023816.2 | NP_076305.2 | 17 | 2910 | ||
| GenBank mRNA | AK005775.1 | - | 2 | 715 | ||
| AK132751.1 | - | 17 | 2909 | |||