siRNA Details
| Gene Symbol: 4930455F16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005958.1 | - | 2 | 280 | ||
| AK015463.1 | - | 2 | 260 | |||
| BU962182.1 | - | 3 | 335 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930455F16Rik| Gene Symbol: 4930455F16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005958.1 | - | 2 | 280 | ||
| AK015463.1 | - | 2 | 260 | |||
| BU962182.1 | - | 3 | 335 | |||