siRNA Details
| Gene Symbol: Il31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029594.1 | NP_083870.1 | 2 | 182 | ||
| GenBank mRNA | AK005939.1 | - | 2 | 352 | ||
| AY509149.1 | - | 2 | 182 | |||
| BC117082.1 | - | 2 | 181 | 40126401 | ||
| BC117084.1 | - | 2 | 181 | 40126403 | ||

Il31
Genomic Map