siRNA Details
| Gene Symbol: Abcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029600.3 | NP_083876.3 | 9 | 1206 | ||
| GenBank mRNA | AF534127.1 | - | 9 | 1206 | ||
| AF534128.1 | - | 9 | 1206 | |||
| AF534129.1 | - | 9 | 1206 | |||
| AK149551.1 | - | 9 | 1206 | |||
| AK155664.1 | - | 9 | 1181 | |||
| AK156615.1 | - | 9 | 1436 | |||
| AK172420.1 | - | 9 | 1436 | |||
| AY795569.1 | - | 9 | 1206 | |||
| AY841885.1 | - | 9 | 1187 | |||
| BC046560.1 | - | 6 | 710 | |||
| BC048825.1 | - | 9 | 1138 | |||
| BC150788.1 | - | 9 | 1197 | 9087699 | ||

Abcc3
Genomic Map