siRNA Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199843.1 | NP_001186772.1 | 2 | 134 | ||
| NM_029601.3 | NP_083877.1 | 2 | 134 | |||
| GenBank mRNA | AK006115.1 | - | 2 | 132 | ||
| BC034668.1 | - | 2 | 72 | |||
| BC038915.1 | - | 2 | 123 | 5148723 | ||
| CF751010.1 | - | 2 | 97 | |||

Ift43
Genomic Map