siRNA Details
| Gene Symbol: Smkr-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006282.1 | - | 2 | 375 | ||
| AK076883.1 | - | 2 | 392 | |||
| BB615595.1 | - | 2 | 389 | |||
| BY706362.1 | - | 2 | 369 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smkr-ps| Gene Symbol: Smkr-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006282.1 | - | 2 | 375 | ||
| AK076883.1 | - | 2 | 392 | |||
| BB615595.1 | - | 2 | 389 | |||
| BY706362.1 | - | 2 | 369 | |||