siRNA Details
| Gene Symbol: Rftn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181397.2 | NP_852062.1 | 7 | 1386 | ||
| GenBank mRNA | AK082517.1 | - | 999 | |||
| AK087803.1 | - | 1160 | ||||
| AK088466.1 | - | 5 | 923 | |||
| AK152130.1 | - | 7 | 1308 | |||
| AK153288.1 | - | 7 | 1308 | |||
| AK155870.1 | - | 7 | 1350 | |||
| AK170913.1 | - | 7 | 1349 | |||
| AK172884.1 | - | 7 | 1314 | |||
| BC043324.1 | - | 3 | 443 | |||
| BC052074.1 | - | 7 | 1386 | 6401192 | ||

Rftn1
Genomic Map