siRNA Details
| Gene Symbol: Daam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008231.2 | NP_001008232.2 | 6 | 639 | ||
| GenBank mRNA | AK122266.1 | - | 6 | 639 | ||
| AY426536.1 | - | 6 | 668 | |||
| BC050043.1 | - | 6 | 639 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Daam2| Gene Symbol: Daam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008231.2 | NP_001008232.2 | 6 | 639 | ||
| GenBank mRNA | AK122266.1 | - | 6 | 639 | ||
| AY426536.1 | - | 6 | 668 | |||
| BC050043.1 | - | 6 | 639 | |||