siRNA Details
| Gene Symbol: Ppp1r18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146710.1 | NP_001140182.1 | 2 | 351 | ||
| NM_001146711.1 | NP_001140183.1 | 2 | 350 | |||
| NM_175242.1 | NP_780451.1 | 1 | 865 | |||
| GenBank mRNA | AK051656.1 | - | 1 | 821 | ||
| AK149943.1 | - | 2 | 352 | |||
| BC020483.1 | - | 2 | 423 | |||
| BC025573.1 | - | 2 | 363 | |||
| BC145183.1 | - | 2 | 320 | 9053734 | ||
| BC150725.1 | - | 2 | 320 | 9087636 | ||

Ppp1r18
Genomic Map