siRNA Details
| Gene Symbol: Ppp1r18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146710.1 | NP_001140182.1 | 2 | 486 | ||
| NM_001146711.1 | NP_001140183.1 | 2 | 485 | |||
| NM_175242.1 | NP_780451.1 | 1 | 1000 | |||
| GenBank mRNA | AK051656.1 | - | 1 | 956 | ||
| AK149943.1 | - | 2 | 487 | |||
| BC020483.1 | - | 2 | 558 | |||
| BC025573.1 | - | 2 | 498 | |||
| BC145183.1 | - | 2 | 455 | 9053734 | ||
| BC150725.1 | - | 2 | 455 | 9087636 | ||

Ppp1r18
Genomic Map