siRNA Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133765.4 | NP_598526.3 | 5 | 696 | ||
| GenBank mRNA | AB041586.1 | - | 5 | 545 | ||
| AK009859.1 | - | 2 | 137 | |||
| AK140412.1 | - | 5 | 697 | |||
| AK163641.1 | - | 5 | 665 | |||
| AK197038.1 | - | 2 | 145 | |||
| BC026929.1 | - | 5 | 681 | 4194857 | ||

Fbxo31
Genomic Map