siRNA Details
| Gene Symbol: Ip6k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029634.2 | NP_083910.2 | 4, 5 | 867 | ||
| GenBank mRNA | AK050593.1 | - | 4, 5 | 723 | ||
| BC039922.1 | - | 4, 5 | 878 | 2609803 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ip6k2| Gene Symbol: Ip6k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029634.2 | NP_083910.2 | 4, 5 | 867 | ||
| GenBank mRNA | AK050593.1 | - | 4, 5 | 723 | ||
| BC039922.1 | - | 4, 5 | 878 | 2609803 | ||