siRNA Details
| Gene Symbol: 1500009C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005178.1 | - | 2 | 870 | ||
| BC023061.1 | - | 2 | 860 | |||
| BC059784.1 | - | 2 | 958 | |||
| BC094356.1 | - | 2 | 806 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500009C09Rik| Gene Symbol: 1500009C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005178.1 | - | 2 | 870 | ||
| BC023061.1 | - | 2 | 860 | |||
| BC059784.1 | - | 2 | 958 | |||
| BC094356.1 | - | 2 | 806 | |||