siRNA Details
| Gene Symbol: Cln6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033175.2 | NP_001028347.1 | 7 | 960 | ||
| GenBank mRNA | AK154414.1 | - | 7 | 961 | ||
| AK154551.1 | - | 7 | 954 | |||
| AK177422.1 | - | 3 | 400 | |||
| BC116794.1 | - | 6 | 786 | |||
| BC116798.1 | - | 6 | 786 | |||
| BC144722.1 | - | 6 | 845 | |||

Cln6
Genomic Map