siRNA Details
| Gene Symbol: Fam204a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029648.6 | NP_083924.2 | 2 | 541 | ||
| GenBank mRNA | AK075914.1 | - | 2 | 542 | ||
| AK162079.1 | - | 2 | 493 | |||
| BC048505.1 | - | 3 | 168 | |||
| BC061459.1 | - | 340 | 6829614 | |||

Fam204a
Genomic Map