siRNA Details
| Gene Symbol: Atg2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029654.4 | NP_083930.5 | 14, 15 | 2579 | ||
| GenBank mRNA | AK085790.1 | - | 14, 15 | 2525 | ||
| BC062182.1 | - | 14, 15 | 2549 | 6505402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atg2b| Gene Symbol: Atg2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029654.4 | NP_083930.5 | 14, 15 | 2579 | ||
| GenBank mRNA | AK085790.1 | - | 14, 15 | 2525 | ||
| BC062182.1 | - | 14, 15 | 2549 | 6505402 | ||