siRNA Details
| Gene Symbol: Styxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289554.1 | NP_001276483.1 | 6 | 708 | ||
| NM_001289555.1 | NP_001276484.1 | 6 | 705 | |||
| NM_001289556.1 | NP_001276485.1 | 5 | 549 | |||
| NM_029659.4 | NP_083935.3 | 6 | 715 | |||
| GenBank mRNA | AK005599.1 | - | 6 | 706 | ||
| AK005858.1 | - | 5 | 456 | |||
| AK161310.1 | - | 6 | 716 | |||
| BC045148.1 | - | 6 | 645 | 6307739 | ||

Styxl1
Genomic Map