siRNA Details
| Gene Symbol: Rbmxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029660.2 | NP_083936.1 | 1 | 1414 | ||
| GenBank mRNA | AK005907.1 | - | 1 | 1415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbmxl2| Gene Symbol: Rbmxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029660.2 | NP_083936.1 | 1 | 1414 | ||
| GenBank mRNA | AK005907.1 | - | 1 | 1415 | ||