siRNA Details
| Gene Symbol: Mfsd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029662.2 | NP_083938.2 | 4, 5 | 651 | ||
| GenBank mRNA | AK006096.1 | - | 4, 5 | 650 | ||
| BC046793.1 | - | 4, 5 | 633 | 5717701 | ||
| BC060526.1 | - | 4, 5 | 622 | 5707105 | ||

Mfsd2a
Genomic Map