siRNA Details
| Gene Symbol: NEK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146099.1 | NP_001139571.1 | 8, 9 | 982 | ||
| NM_002498.2 | NP_002489.1 | 8, 9 | 982 | |||
| NM_152720.2 | NP_689933.1 | 8, 9 | 1001 | |||
| NR_027415.2 | 8, 9 | 982 | ||||
| GenBank mRNA | AB072828.1 | - | 7, 8 | 651 | ||
| AK131359.1 | - | 7, 8 | 1900 | |||
| AK290259.1 | - | 8, 9 | 714 | |||
| BC019916.1 | - | 8, 9 | 709 | 5000918 | ||
| CV029372.1 | - | 7, 8 | 606 | |||
| DA407862.1 | - | 2, 3 | 137 | |||
| Z29067.1 | - | 6, 7 | 465 | |||

NEK3
Genomic Map