siRNA Details
| Gene Symbol: Mib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256107.1 | NP_001243036.1 | 14 | 2654 | ||
| NM_001256108.2 | NP_001243037.2 | 14 | 2474 | |||
| NM_145124.3 | NP_660106.2 | 13 | 2558 | |||
| GenBank mRNA | AB072336.1 | - | 13 | 2120 | ||
| AK008671.1 | - | 1 | 48 | |||
| AK084791.1 | - | 6 | 856 | |||
| AK147940.1 | - | 14 | 2419 | |||
| AK159611.1 | - | 14 | 2371 | |||
| AY974090.1 | - | 12 | 1855 | |||
| BC058086.1 | - | 11 | 2142 | |||
| BC141133.1 | - | 11 | 1947 | 9055745 | ||
| BC145441.1 | - | 12 | 2043 | |||

Mib2
Genomic Map