siRNA Details
| Gene Symbol: Mib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256107.1 | NP_001243036.1 | 6 | 1487 | ||
| NM_001256108.2 | NP_001243037.2 | 7 | 1403 | |||
| NM_145124.3 | NP_660106.2 | 6 | 1487 | |||
| GenBank mRNA | AB063290.1 | - | 4 | 492 | ||
| AK147940.1 | - | 7 | 1347 | |||
| AK159611.1 | - | 6 | 1205 | |||
| AY974090.1 | - | 4 | 628 | |||
| BC058086.1 | - | 5 | 746 | |||
| BC141133.1 | - | 5 | 801 | 9055745 | ||
| BC145441.1 | - | 5 | 801 | |||

Mib2
Genomic Map