siRNA Details
| Gene Symbol: 1700112E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028275.1 | NP_082551.1 | 4 | 479 | ||
| GenBank mRNA | AK007175.1 | - | 4 | 483 | ||
| AK012226.1 | - | 2 | 304 | |||
| AK153491.1 | - | 3 | 344 | |||
| BY516672.1 | - | 24 | ||||

1700112E06Rik
Genomic Map