siRNA Details
| Gene Symbol: 1700113H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029685.1 | NP_083961.1 | 6 | 828 | ||
| GenBank mRNA | AK007195.1 | - | 2 | 299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700113H08Rik| Gene Symbol: 1700113H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029685.1 | NP_083961.1 | 6 | 828 | ||
| GenBank mRNA | AK007195.1 | - | 2 | 299 | ||