siRNA Details
| Gene Symbol: Pkd1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029686.4 | NP_083962.4 | 41 | 7019 | ||
| GenBank mRNA | AY164484.1 | - | 41 | 7019 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkd1l2| Gene Symbol: Pkd1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029686.4 | NP_083962.4 | 41 | 7019 | ||
| GenBank mRNA | AY164484.1 | - | 41 | 7019 | ||