siRNA Details
| Gene Symbol: Srxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029688.5 | NP_083964.2 | 2 | 1509 | ||
| GenBank mRNA | AK089976.1 | - | 2 | 1306 | ||
| AK159226.1 | - | 2 | 1304 | |||
| AK159613.1 | - | 2 | 1303 | |||
| AK161066.1 | - | 2 | 1346 | |||
| BC049957.1 | - | 2 | 1509 | 4507800 | ||

Srxn1
Genomic Map