siRNA Details
| Gene Symbol: Srxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029688.5 | NP_083964.2 | 2 | 1204 | ||
| GenBank mRNA | AK007296.1 | - | 2 | 999 | ||
| AK089976.1 | - | 2 | 1001 | |||
| AK159226.1 | - | 2 | 999 | |||
| AK159613.1 | - | 2 | 998 | |||
| AK161066.1 | - | 2 | 1041 | |||
| BC011325.1 | - | 2 | 1018 | 3154456 | ||
| BC049957.1 | - | 2 | 1204 | 4507800 | ||

Srxn1
Genomic Map