siRNA Details
| Gene Symbol: Cfap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163638.1 | NP_001157110.1 | 2, 3 | 182 | ||
| NM_001163639.1 | NP_001157111.1 | 2, 3 | 182 | |||
| NM_029698.1 | NP_083974.1 | 2, 3 | 120 | |||
| GenBank mRNA | AK017232.1 | - | 2, 3 | 183 | ||
| AK029536.1 | - | 2, 3 | 118 | |||
| BC138269.1 | - | 2, 3 | 179 | 8861288 | ||
| BC145225.1 | - | 2, 3 | 179 | 9053777 | ||
| BC145226.1 | - | 2, 3 | 179 | 9053778 | ||

Cfap70
Genomic Map