siRNA Details
| Gene Symbol: Trim12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023835.2 | NP_076324.2 | 2 | 284 | ||
| GenBank mRNA | AF220126.1 | - | 2 | 377 | ||
| AK009769.1 | - | 2 | 246 | |||
| BC094899.1 | - | 2 | 244 | 30041200 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim12a| Gene Symbol: Trim12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023835.2 | NP_076324.2 | 2 | 284 | ||
| GenBank mRNA | AF220126.1 | - | 2 | 377 | ||
| AK009769.1 | - | 2 | 246 | |||
| BC094899.1 | - | 2 | 244 | 30041200 | ||