siRNA Details
| Gene Symbol: Spcs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029701.1 | NP_083977.1 | 3 | 345 | ||
| GenBank mRNA | AK007432.1 | - | 3 | 324 | ||
| AK028314.1 | - | 3 | 307 | |||
| AK048051.1 | - | 3 | 331 | |||
| BC019390.1 | - | 3 | 284 | |||
| BC054817.1 | - | 3 | 310 | 5375997 | ||
| BY012914.1 | - | 3 | 345 | |||

Spcs3
Genomic Map