siRNA Details
| Gene Symbol: Trdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029726.2 | NP_084002.2 | 2 | 287 | ||
| GenBank mRNA | AF223415.1 | - | 2 | 230 | ||
| AF223416.1 | - | 2 | 230 | |||
| AF223417.1 | - | 2 | 209 | |||
| AK009816.1 | - | 2 | 260 | |||
| BB658668.1 | - | 2 | 289 | |||
| BC034343.1 | - | 2 | 210 | 3589077 | ||

Trdn
Genomic Map