siRNA Details
| Gene Symbol: Gsdma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029727.2 | NP_084003.2 | 6 | 766 | ||
| GenBank mRNA | AK008613.1 | - | 4 | 419 | ||
| AK008663.1 | - | 6 | 763 | |||
| BC109335.1 | - | 5 | 669 | 40052139 | ||
| BC109336.1 | - | 6 | 700 | 40052146 | ||

Gsdma2
Genomic Map