siRNA Details
| Gene Symbol: Gsdma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029727.2 | NP_084003.2 | 5 | 638 | ||
| GenBank mRNA | AK008663.1 | - | 5 | 635 | ||
| BC109335.1 | - | 4 | 541 | 40052139 | ||
| BC109336.1 | - | 5 | 572 | 40052146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsdma2| Gene Symbol: Gsdma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029727.2 | NP_084003.2 | 5 | 638 | ||
| GenBank mRNA | AK008663.1 | - | 5 | 635 | ||
| BC109335.1 | - | 4 | 541 | 40052139 | ||
| BC109336.1 | - | 5 | 572 | 40052146 | ||