siRNA Details
| Gene Symbol: 2010005H15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029733.3 | NP_084009.1 | 3, 4 | 244 | ||
| GenBank mRNA | AK008122.1 | - | 3, 4 | 339 | ||
| BC125396.1 | - | 3, 4 | 244 | 40129911 | ||
| BC125422.1 | - | 3, 4 | 244 | 40129937 | ||
| BC145264.1 | - | 2, 3 | 182 | 9053817 | ||

2010005H15Rik
Genomic Map